Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD11b FITC, Clone: M1/70,
Rat Monoclonal Antibody
$189.29 - $366.62
Specifications
Antigen | CD11b |
---|---|
Clone | M1/70 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Description
Specifically detects CD11b Clone: M1/70 in Human, Mouse samples, and it is validated for Flow Cytometry
The M1/70 antibody reacts with human and mouse CD11b, also known as integrin alpha M. This 165-170kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alphaM (CD11b) assembles with Integrin beta-2 (CD18) into a receptor known as Macrophage Antigen-1 (Mac-1) or complement receptor type 3 (CR3). This receptor binds and induces intracellular signaling through ICAM-1 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.The M1/70 antibody is widely used as a marker for CD11b expression on mouse macrophages, granulocytes, neutrophils, and NK cells. The antibody is also reported to be cross-reactive for Rhesus macaque CD11b.Specifications
CD11b | |
Flow Cytometry | |
FITC | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Itgam | |
IgG2b κ | |
Purified by size-exclusion chromatography |
M1/70 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
P05555 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title