Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD11c PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
$184.44 - $393.24
Specifications
Antigen | CD11c |
---|---|
Clone | ICRF 3.9 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1777P025
|
R&D Systems
FAB1777P025 |
25 Tests |
Each for $184.44
|
|
|||||
FAB1777P100
|
R&D Systems
FAB1777P100 |
100 Tests |
Each for $393.24
|
|
|||||
Description
CD11c Monoclonal specifically detects CD11c in Human samples. It is validated for Flow Cytometry.Specifications
CD11c | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
3687 | |
Rheumatoid synovial fluid cells and human monocyte-derived fibronectin | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
ICRF 3.9 | |
Flow Cytometry | |
R-PE | |
Mouse | |
Human | |
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, Integrin alpha X, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), ITGAX, Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, p150,95 alpha, SLEB6 | |
ITGAX | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human CD11c in flow cytometry. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title