Learn More
CD123 Mouse anti-Human, PerCP-Cy5.5, Clone: 7G3, BD
Mouse Monoclonal Antibody
$188.00 - $436.00
Specifications
| Antigen | CD123 |
|---|---|
| Clone | 7G3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PerCP-Cyanine5.5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB560904
|
BD Biosciences
560904 |
25 Tests |
Each for $188.00
|
|
|||||
|
BDB558714
|
BD Biosciences
558714 |
100 Tests |
Each for $436.00
|
|
|||||
Description
Reacts with human CD123, the 70 kD IL-3 receptor α chain (IL-3Rα), which associates with the 120-140kDa β subunit. The β chain is shared with the receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on hematopoietic progenitors. This antibody has been reported to block the binding of 125I-IL-3 to high and low affinity IL-3 receptors. In functional experiments, this antibody was found to inhibit acute myeloid leukemia cell proliferation, basophil histamine release, endothelial cell-mediated IL-8 secretion, and neutrophil transmigration. This antibody has been reported to be useful for immunoprecipitation, western blot and immunofluorescent staining for flow cytometry. IL-3Rα plays an important role in hematopoietic progenitor cell growth and differentiation. At the Fifth HLDA Workshop, the human IL-3 receptor was designated CD123.
Flow Cytometry
Specifications
| CD123 | |
| Flow Cytometry | |
| PerCP-Cyanine5.5 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 7G3 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human IL-3Ra-transfected cells | |
| Primary | |
| IL3RA; IL-3RA; IL-3Rα; IL-3R-alpha; Interleukin-3 receptor subunit alpha |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.