Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158b Mouse anti-Human, BB515, Clone: CH-L, BD

Catalog No. BDB564678 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564678 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564678 Supplier BD Biosciences Supplier No. 564678

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Mouse Monoclonal Antibody

The CH-L monoclonal antibody specifically binds to CD158b proteins. These proteins are 50-58 kDa type I glycoproteins that belong to the Killer cell immunoglobulin-like receptor (KIR) family. They are also known as CD158b1 (KIR2DL2; NKAT-6; p58.2), CD158b2 (KIR2DL3; NKAT-2; p58.2), or CD158j (KIR2DS2; NKAT-5; p50.2). The CD158b molecules are composed of two extracellular Ig-like domains, and a transmembrane region. CD158b1 and CD158b2 also possess long (84 or 76 amino acids, respectively) cytoplasmic tails with two immunoreceptor tyrosine-based inhibition motifs (ITIM) whereas CD158j has a short (39 amino acid) cytoplasmic tail that lacks the ITIM motif. CD158b molecules are expressed on NK cells and subsets of TCR αβ+ cells or TCR γδ+ cells. Ligand- or CH- L antibody-bound CD158b1 or CD158b2 can reportedly inhibit cytolytic NK and T cell responses to various stimuli including certain target cells expressing MHC class I ligands encoded by HLA-C alleles (Cw 1, 3, 7 and 8). CD158j reportedly can enhance some cellular cytolytic responses.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158b
Applications Flow Cytometry
Classification Monoclonal
Clone CH-L
Conjugate Brilliant Blue 515 (BB515)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.