Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD201 Rat anti-Human, APC, Clone: RCR-252, BD

Catalog No. BDB563622 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563622 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563622 Supplier BD Biosciences Supplier No. 563622
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The RCR-252 monoclonal antibody specifically binds to the endothelial cell protein C receptor (EPCR), which is also known as CD201. CD201 is a 25 kDa, type I transmembrane glycoprotein, expressed on endothelial cells in arteries, veins and capillaries. CD201 binds activated protein C, a serine protease that is involved in regulating blood coagulation. EPCR is reportedly expressed on several malignant cell lines. Malignant cell lines that express EPCR can show significant levels of protein C activation and this activation can be inhibited by anti-EPCR antibody

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD201
Applications Flow Cytometry
Classification Monoclonal
Clone RCR-252
Conjugate APC
Description EPCR; Activated protein C receptor; APC receptor; PROCR; centrocyclin
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen EPCR-positive RE-1 Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.