Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD205 Mouse anti-Human, PE, Clone: MG38, BD

Catalog No. BDB558069 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558069 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558069 Supplier BD Biosciences Supplier No. 558069
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with DEC-205, a 205 kDa transmembrane protein with C-type lectin external domains, present on dendritic cells (DC) and some epithelia. It is being described as a multilectin receptor for adsorptive endocytosis. MG38 antibody reacted with mature monocyte-derived DC and also with B cells, monocytes and weakly on T cells and NK cells. Antibody MG38 is useful for the study of DC development.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD205
Applications Flow Cytometry
Classification Monoclonal
Clone MG38
Conjugate PE
Description DEC-205; Lymphocyte Antigen 75; CLEC13B
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.