Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Mouse anti-Human, FITC, Clone: L128, BD

Catalog No. BDB340424 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340424 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340424 Supplier BD Biosciences Supplier No. 340424
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD27 antibody, clone L128, is derived from hybridization of mouse Sp2/0 cells with spleen cells from BALB/c mice immunized with activated peripheral blood lymphocytes.

The CD27 antibody recognizes a 110–120-kilodalton (kDa) disulfide-linked homodimer comprised of two 55-kDa polypeptide chains. The CD27 antigen is a lymphocyte-specific member of the tumor necrosis factor receptor (TNFR) super family, which also includes nerve growth factor receptor, CD30, CD40, CD95 (Fas), CD120a, CD120b, CD134 (OX 40), and CD137. The CD27 antigen is also known as S152, T14, TNFRSF7, and Tp55.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD27
Applications Flow Cytometry
Classification Monoclonal
Clone L128
Concentration 0.8μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Activated Peripheral Blood Cells
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.