Learn More
CD3 Mouse anti-Human, BB515, Clone: UCHT1, BD
Mouse Monoclonal Antibody
$70.30 - $144.60
Specifications
| Antigen | CD3 |
|---|---|
| Clone | UCHT1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
Description
The UCHT1 monoclonal antibody specifically binds to the human CD3 ε -chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3 ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3 ε -positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3 ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3 ε .
The antibody was conjugated to BD Horizon™ BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Specifications
| CD3 | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| UCHT1 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.