Learn More
CD32 Mouse anti-Human, Unlabeled, Clone: FLI8.26, BD
Mouse Monoclonal Antibody
$182.00
Specifications
Antigen | CD32 |
---|---|
Clone | FLI8.26 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Reacts with the 40kDa type I membrane glycoprotein, FcγRII, the low affinity receptor for aggregated IgG expressed on B cells, monocytes, granulocytes, and platelets. CD32 mediates endocytosis, cytotoxicity, platelet activation and immunomodulation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: FLI8.26 (2003)
Isotype: IgG2b, κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Specifications
CD32 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C |
FLI8.26 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
K562 or FcγRII+ L cell lines | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.