Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD4 FITC, Clone: RM4-5,
Rat Monoclonal Antibody
$115.17 - $189.28
Specifications
Antigen | CD4 |
---|---|
Clone | RM4-5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1567MI
|
MilliporeSigma™
MABF1567 |
25 μg |
Each for $189.28
|
|
|||||
MABF1568MI
|
MilliporeSigma™
MABF1568 |
100 μg |
Each for $115.17
|
|
|||||
Description
Specifically detects CD4 Clone: RM4-5 in Mouse samples, and it is validated for Flow Cytometry
The RM4-5 antibody reacts with mouse CD4, a 55kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta2-domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.The RM4-5 antibody is widely used as a phenotypic marker for CD4 expression. If used together, the RM4-5 antibody and an alternative antibody, Anti-Mouse CD4 clone GK1.5, will 'compete' for binding, i.e. RM4-5 antibody is able to block GK1.5 antibody binding to cells. In contrast, RM4-5 antibody does not block the binding of Anti-Mouse CD4 clone RM4-4 to cells.Specifications
CD4 | |
Flow Cytometry | |
FITC | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Cd4 | |
IgG2a κ | |
Purified by size-exclusion chromatography |
RM4-5 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P06332 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title