Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD44 Violer Fluor 710, Clone: IM7,
Rat Monoclonal Antibody
$189.29 - $338.58
Specifications
Antigen | CD44 |
---|---|
Clone | IM7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Violet Fluor 710 (V710) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1563MI
|
MilliporeSigma™
MABF1563 |
25 μg |
Each for $189.29
|
|
|||||
MABF1564MI
|
MilliporeSigma™
MABF1564 |
100 μg |
Each for $338.58
|
|
|||||
Description
Specifically detects CD44 Clone: IM7 in Human, Mouse samples, and it is validated for Flow Cytometry
The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or 'standard' form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141).Specifications
CD44 | |
Flow Cytometry | |
Violet Fluor 710 (V710) | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Cd44, Ly-24 | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
IM7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
P15379 | |
Proprietary immunogen | |
Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title