Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD45, PE-Cy7, Clone: 30-F11,
Rat Monoclonal Antibody
$189.29 - $277.47
Specifications
Antigen | CD45 |
---|---|
Clone | 30-F11 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1464MI
|
MilliporeSigma™
MABF1464 |
25 μg |
Each for $189.29
|
|
|||||
MABF1465MI
|
MilliporeSigma™
MABF1465 |
100 μg |
Each for $277.47
|
|
|||||
Description
Specifically detects CD45 Clone: 30-F11 in Mouse samples, and it is validated for Flow Cytometry
The 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others. The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.Specifications
CD45 | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Ptprc, Ly-5 | |
IgG2b κ | |
Purified by size-exclusion chromatography |
30-F11 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P06800 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title