Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49F Rat anti-Human, Brilliant Violet 650, Clone: GOH3, BD

Rat Monoclonal Antibody

Supplier:  BD Biosciences 563707

 View more versions of this product

Catalog No. BDB563707


Only null left
Add to Cart

Description

Description

The GoH3 monoclonal antibody specifically binds to CD49f which is also known as i ntegrin α6 chain . CD49f is a ∽150 kDa type I transmembrane glycoprotein that belongs to the integrin alpha chain family of extracellular matrix and cell adhesion receptors. The i ntegrin α6 subunit associates with the i ntegrin β1 chain (CD29) to form VLA-6 and with the integrin β4 chain (CD104) to form the integrin α6β4 complex, also known as the laminin and kalinin receptor. CD49f is ex pressed mainly on T cells, monocytes, platelets, epithelial cells, endothelial cells, perineural cells, and trophoblasts of placenta. GoH3 recognizes an extracellular epitope of integrin α6 on human, mouse and bovine cells. GoH3 has been reported to block the binding of integrin α6 to laminin P1 and E8 fragments.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD49f
Monoclonal
Brilliant Violet 650
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse mammary tumor cells
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
GOH3
ITGA6; IThe GoH3 monoclonal ntegrin alpha-6; Integrin α6 chain; VLA-6; ITA6
Rat
Affinity Purified
RUO
Canine, Human, Mouse, Pig
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.