Learn More
Description
The migration of T cells through lymph nodes and their interactions with antigen presenting cells (APCs) involve adhesion molecules on the surfaces of the T cell and the cell with which it interacts. These molecules include the selectins, the integrins, members of the immunoglobulin superfamily, and some mucin-like proteins. The integrins mediate adhesion between cells and between cells and the extracellular matrix. LFA-1 (lymphocyte function-associated antigen 1), a member of the β2 integrin family, is expressed on T cells, neutrophils, and macrophages and is thought to be the most important adhesion molecule for lymphocyte activation. It interacts with members of the immunoglobulin superfamily, the intercellular adhesion molecules: ICAM-1, -2, and -3. ICAM-1 and ICAM-2 are expressed on endodthelium and APCs, while ICAM-3 is expressed only on leukocytes. ICAM-1 is most homologous to neural cell adhesion molecule and myelin-associated glycoprotein. It is a 55kDa glycoprotein that is heavily glycosylated to form 90kDa to 115kDa proteins. Its interactions with LFA-1 are necessary for CTL and NK mediated killing, T and B cell responses, and immune cell extravasation.
Host Species: Mouse
Clone: 28
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human CD54/ICAM-1 aa. 46-160
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | CD54 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 28 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 150 μg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.