Learn More
CD54 (ICAM-1) Mouse anti-Human, Unlabeled, Clone: 28, BD
Mouse Monoclonal Antibody
$310.50
Specifications
| Antigen | CD54 |
|---|---|
| Clone | 28 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB611704
|
BD Biosciences
611704 |
50 μg |
Each for $310.50
|
|
|||||
|
BDB611705
|
BD Biosciences
611705 |
150 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
The migration of T cells through lymph nodes and their interactions with antigen presenting cells (APCs) involve adhesion molecules on the surfaces of the T cell and the cell with which it interacts. These molecules include the selectins, the integrins, members of the immunoglobulin superfamily, and some mucin-like proteins. The integrins mediate adhesion between cells and between cells and the extracellular matrix. LFA-1 (lymphocyte function-associated antigen 1), a member of the β2 integrin family, is expressed on T cells, neutrophils, and macrophages and is thought to be the most important adhesion molecule for lymphocyte activation. It interacts with members of the immunoglobulin superfamily, the intercellular adhesion molecules: ICAM-1, -2, and -3. ICAM-1 and ICAM-2 are expressed on endodthelium and APCs, while ICAM-3 is expressed only on leukocytes. ICAM-1 is most homologous to neural cell adhesion molecule and myelin-associated glycoprotein. It is a 55kDa glycoprotein that is heavily glycosylated to form 90kDa to 115kDa proteins. Its interactions with LFA-1 are necessary for CTL and NK mediated killing, T and B cell responses, and immune cell extravasation.
Host Species: Mouse
Clone: 28
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human CD54/ICAM-1 aa. 46-160
Immunofluorescence, Western Blotting
Specifications
| CD54 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| IgG1 | |
| Affinity Purified |
| 28 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.