Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Mouse anti-Human, FITC, Clone: M-B741, BD

Catalog No. BDB555540 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555540 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555540 Supplier BD Biosciences Supplier No. 555540
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with cell -surface MHC class II-associated invariant chain molecule (41, 35, 33 kD). CD74 is moderately expressed on B cells, weakly on monocytes and macrophages. Other reactive cells include Langerhans cells, dendritic cells, a subset of activated T cells and thymic epithelium. CD74 is postulated to play a role in antigen processing and presentation through intracellular transport and expression of class II molecules on the cell surface.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD74
Applications Flow Cytometry
Classification Monoclonal
Clone M-B741
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.