Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 Mouse anti-Human, BB515, Clone: L307.4, BD

Catalog No. BDB565009 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565009 25 Tests
BDB565008 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565009 Supplier BD Biosciences Supplier No. 565009
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L307.4 monoclonal antibody specifically binds to B7/BB1, a 60 kDa transmembrane glycoprotein that was clustered as CD80 in the Fifth International Workshop on Human Leukocyte Differentiation Antigens. CD80, a member of the Ig supergene family, is expressed on activated B cells, T cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells, CD28 and CD152 (CTLA-4). CD80 is also expressed on activated CD4-positive and CD8-positive T cells, appearing late after activation suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 by anti-CD28 or by CD80 results in T-cell activation and a signal for IL-2 production.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone L307.4
Conjugate Brilliant Blue 515 (BB515)
Description B7.1; B7-1; Activation B7-1 antigen; B7; BB1; CD28LG; CD28LG1; LAB7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide
Host Species Mouse
Immunogen Human B7-transfected L cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.