Learn More
CD80 Mouse anti-Human, V450, Clone: L307.4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560444
Description
Reacts with B7/BB1, a 60kDa transmembrane glycoprotein which was clustered as CD80 in the Fifth International Workshop on Human Leukocyte Differentiation Antigens. CD80, a member of the Ig supergene family, is expressed on activated B cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells, CD28 and CD152 (CTLA-4). CD80 is also expressed on activated CD4+ and CD8+ T cells, appearing late after activation suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 by anti-CD28 or by CD80 results in T-cell activation and a signal for IL-2 production. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: L307.4
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD80 | |
Monoclonal | |
Violet Fluor 450 (V450) | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human B7-transfected L cells | |
30 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
L307.4 | |
B7.1; B7-1; Activation B7-1 antigen; B7; BB1; CD28LG; CD28LG1; LAB7 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.