Learn More
CD80 Mouse anti-Human, V450, Clone: L307.4, BD
Mouse Monoclonal Antibody
$182.00 - $426.00
Specifications
Antigen | CD80 |
---|---|
Clone | L307.4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Violet Fluor 450 (V450) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560444
|
BD Biosciences
560444 |
30 Tests |
Each for $182.00
|
|
|||||
BDB560442
|
BD Biosciences
560442 |
120 Tests |
Each for $426.00
|
|
|||||
Description
Reacts with B7/BB1, a 60kDa transmembrane glycoprotein which was clustered as CD80 in the Fifth International Workshop on Human Leukocyte Differentiation Antigens. CD80, a member of the Ig supergene family, is expressed on activated B cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells, CD28 and CD152 (CTLA-4). CD80 is also expressed on activated CD4+ and CD8+ T cells, appearing late after activation suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 by anti-CD28 or by CD80 results in T-cell activation and a signal for IL-2 production. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: L307.4
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
CD80 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
Human | |
Human B7-transfected L cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
L307.4 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
B7.1; B7-1; Activation B7-1 antigen; B7; BB1; CD28LG; CD28LG1; LAB7 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.