Learn More
CD95 Mouse, Unlabeled, Clone: 13, BD
Mouse Monoclonal Antibody
Specifications
Antigen | CD95 |
---|---|
Clone | 13 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB610197
|
BD Biosciences
610197 |
50 μg | N/A | N/A | N/A | ||||
Description
CD95 is a member of a family of cell surface receptors that includes tumor necrosis factor receptor (TNF-R), nerve growth factor receptor (NGF-R), CD40, CD27, CD30, and 4-1BB. Both murine and human Fas genes have been cloned and reportedly share 60% similarity in their amino acid sequences. CD95 (Fas) is a cell surface apoptosis-signaling molecule that is widely expressed in sites such as thymus, liver, heart, and ovary. Abnormalities in the Fas gene correlate with autoimmune features in mice and with unusually high levels of lymphocyte apoptosis in HIV-infected humans. Genetic studies have localized the Fas gene near the lpr (lymphoproliferation disease) locus on mouse chromosome 19 and further characterization reportedly has revealed that lpr is a mutation affecting the function of the Fas gene.
Specifications
CD95 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Fas, APO-1 |
13 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.