Learn More
MilliporeSigma™ anti-Choline Acetyltransferase Ab-4; Polyclonal
Supplier:  MilliporeSigma™ AB144P200UL/DEL
 View more versions of this product
Catalog No. 50-173-063
Description
Description
Acetylcholine (ACh) is a common neurotransmitter for motoneurons, preganglionic autonomic neurons, postganglionic parasympathetic neurons, a variety of brain regions and some emerging neuron-like stem cells. The metabolism of Ach is relatively simple, involving only two enzymes: choline acetyltransferase (ChAT) for synthesis and acetylcholinesterase (AChE) for degradation. Further, acetylcholine has little function in neurons other than neurotransmission and seems to be neuron specific. It seems that only cholinergic neurons have significant amounts of ChAT making anti-cholinacetyltransferase a useful specific marker.
Immunohistochemistry, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Choline Acetyltransferase Ab-4
Unconjugated
Human placental enzyme.
200 μL
Neuroscience
Avian, Chicken, Guinea Pig, Human, Mouse, Rat
Polyclonal
Goat
Affinity Purified
RUO
Primary
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.