Learn More
MilliporeSigma™ anti-Choline Acetyltransferase Ab-4; Polyclonal
Specifications
Antigen | Choline Acetyltransferase Ab-4 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Goat |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-059
|
MilliporeSigma™
AB144/DEL |
50 μL | N/A | N/A | N/A | ||||
50-173-063
|
MilliporeSigma™
AB144P200UL/DEL |
200 μL | N/A | N/A | N/A | ||||
50-173-061
|
MilliporeSigma™
AB144P/DEL |
500 μL | N/A | N/A | N/A | ||||
50-173-062
|
MilliporeSigma™
AB144P1ML/DEL |
1 mL | N/A | N/A | N/A | ||||
Description
Acetylcholine (ACh) is a common neurotransmitter for motoneurons, preganglionic autonomic neurons, postganglionic parasympathetic neurons, a variety of brain regions and some emerging neuron-like stem cells. The metabolism of Ach is relatively simple, involving only two enzymes: choline acetyltransferase (ChAT) for synthesis and acetylcholinesterase (AChE) for degradation. Further, acetylcholine has little function in neurons other than neurotransmission and seems to be neuron specific. It seems that only cholinergic neurons have significant amounts of ChAT making anti-cholinacetyltransferase a useful specific marker.
Immunohistochemistry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Choline Acetyltransferase Ab-4 | |
Unconjugated | |
RUO | |
Human placental enzyme. |
Polyclonal | |
Goat | |
Neuroscience | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.