Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cleaved-Tau (Asp421) Mouse anti-Human, Mouse, Rat, Clone: C3, MilliporeSigma™

Catalog No. 36017MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-017-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-017-MI Supplier MilliporeSigma Supplier No. 36017
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-cleaved-Tau (Asp421) Antibody, clone C3 is an antibody against cleaved-Tau (Asp421) for use in immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen cleaved-Tau (Asp421)
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C3
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol
Gene Accession No. NM_005910.3; NM_016834.2; NM_016835.2; NM_016841.2
Gene Symbols MAPT, MTBT2, MAPTL, PHF-tau, tau, DDPAC, FTDP-17, MGC138549, MSTD, TAU, FLJ31424, MTBT1, PPND
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 412-421(CSSTGSIDMVD) of human Tau with a Cys at the N-terminal end
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.