Learn More
CLIP-115 Mouse, Unlabeled, Clone: 14, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611143
Description
Microtubules, integral components of the cytoskeleton, are essential for establishment of cell polarity, membrane structure organization, and organelle translocation. Cell polarity is accomplished through the linkage of microtubules to other cytoskeletal proteins by microtubule associated proteins (MAPs). Likewise, the process of organelle translocation requires accessory proteins that link the organelles to microtubules. These proteins are known as CLIPs (cytoplasmic linker proteins) and belong to a novel family of microtubule binding proteins that are characterized by the presence of N-terminal microtubule binding (MTB) domains. CLIP-115 was discovered due to its high degree of homology to CLIP-170 (restin), which links endosomes to microtubules. Like CLIP-170, CLIP-115 contains two MTB domains and a long coiled-coil region. CLIP-115 has been reported to be found primarily in the brain where it associates with microtubules and the dendritic lamellar body (DLB). DLBs have been reported to be organelles that are present exclusively in the bulbous dendritic appendages of neurons. It is thought that CLIP-115 is a protein constituent of DLBs and is responsible for determining their structure and cytoplasmic location.
Host Species: Mouse
Clone: 14
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat CLIP-115 aa. 801-917
Formula Weight [Chemical]: 115kDa
Immunofluorescence, Western Blotting

Specifications
CLIP-115 | |
Monoclonal | |
250μg/mL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
IgG1 |
Immunofluorescence, Western Blot | |
14 | |
Unconjugated | |
Rat CLIP-115 aa. 801-917 | |
150 μg | |
Cell Biology | |
Human, Mouse, Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.