Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CNPase, Clone: 11-5B, ™ (Chemicon™)

Catalog No. 50175222 Shop All MilliporeSigma Products
Change view
Click to view available options
Immunogen:
Purified human 2', 3'-cyclic nucleotide 3'-phosphodiesterase
Purified human brain CNPase.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Immunogen
50-175-222 Purified human brain CNPase.
50-175-225 Purified human 2', 3'-cyclic nucleotide 3'-phosphodiesterase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-175-222 Supplier MilliporeSigma™ Supplier No. MAB326/DEL

CNPase (2', 3'-cyclic nucleotide 3'-phosphodiesterase [or -phosphohydrolase], EC 3.1.4.37) is present in very high levels in brain and peripheral nerve. This enzyme is found almost exclusively in oligo-dendrocytes and Schwann cells, the cells that form myelin in the central and peripheral nervous system, respectively. Therefore, CNPase has been very useful as a marker for these particular cell types. Developmentally, this enzyme appears early in oligodendrocytes, earlier than most other myelin proteins. It continues to be expressed at high levels in oligodendrocytes and Schwann cells of adult animals. In various diseases, neurological mutants, and in experimental conditions in which myelin is reduced, CNPase levels may be severely reduced. The enzyme itself exists in two forms in most species (three in mouse) with molecular masses of 48 and 46 kD on SDS-PAGE, referred to as CNP2 and CNP1, respectively.Studies continue to elucidate the physiological role of the enzyme. Immunohistochemical localization of CNPase has shown the enzyme to be restricted to oligodendrocytes and Schwann cells. The enzyme appears to be distributed in single and double loose wraps of myelin and not in compact myelin as earlier thought by most investigators. CNPase is located on the inner and outer loops of myelin, paranodally and near the inner surface of the oligodendrocyte membrane. In plaque regions of multiple sclerosis patients, the enzyme is reduced, and when CNS myelin is decreased, CNPase is one of the earlier proteins to be lost from the myelin. In addition, an enzyme that is probably identical to brain CNPase is located in the outer rod segments within the visual system, and this protein is also recognized by the monoclonal antibody 11-5B. In mixed human gliomas, the enzyme levels are reduced, although about 5% of the oligodendrocytes occasionally show normal positive staining.

Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

TRUSTED_SUSTAINABILITY

Specifications

Antigen CNPase
Classification Monoclonal
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human brain CNPase.
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Bovine, Canine, Human, Mouse, Pig, Rabbit, Rat, Sheep
Form Semi-purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.