Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3006MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3006MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3006MI Supplier MilliporeSigma Supplier No. AB3006
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-CREB Antibody (Rabbit Polyclonal Antibody) validated in ELISA, EMSA, immunoprecipitation, western blotting to detect CREB also known as CREB-1, cAMP-response Element-binding Protein-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB
Applications Electromobility Shift Assay, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid (sterile filtered) in 0.02M Potassium Phosphate buffer containing 0.15M Sodium Chloride, pH 7.2 with 0.01% sodium azide.
Gene Symbols CREB1; MGC9284; CREB
Host Species Rabbit
Immunogen CREB-1 (p43) peptide corresponding to the N-terminal (aa 5-45) of human protein conjugated to KLH.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_004379.2; NM_134442.2
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.