Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crk Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610035
Description
Crk was first isolated as the v- crk oncogene from chicken retroviruses CT10 and ASV-1. All human cell lines examined to date express a 40 kDa Crk protein. In addition, there is variable expression of 42 kDa and 28 kDa Crk proteins. The c- crk gene is one of a class of genes, such as Nck and GRB2/ASH , which encode proteins that consist mainly of SH2 and SH3 domains. These proteins function as adaptor molecules in tyrosine kinase signal transduction pathways. The SH2 domains interact with phosphotyrosine-containing peptides, while the SH3 domains can enhance this interaction and/or bind to other cellular components. Both the SH2 and SH3 domains of the human Crk protein are required for differentiation of PC12 cells. Thus, Crk has a role in an NGF-induced signaling pathway that involves activation of p21ras. Furthermore, three proteins of 118 kDa, 125 kDa, and 136 kDa which specifically bind to the Crk SH3 domain have been identified.
Specifications
| Crk | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human Crk aa. 102 to 304 | |
| 50 μg | |
| Cell Biology | |
| Bovine, Canine, Chicken, Frog, Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| 22 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction