Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crk Mouse, Unlabeled, Clone: 22, BD
Mouse Monoclonal Antibody
$280.00 - $556.00
Specifications
| Antigen | Crk |
|---|---|
| Clone | 22 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Crk was first isolated as the v- crk oncogene from chicken retroviruses CT10 and ASV-1. All human cell lines examined to date express a 40 kDa Crk protein. In addition, there is variable expression of 42 kDa and 28 kDa Crk proteins. The c- crk gene is one of a class of genes, such as Nck and GRB2/ASH , which encode proteins that consist mainly of SH2 and SH3 domains. These proteins function as adaptor molecules in tyrosine kinase signal transduction pathways. The SH2 domains interact with phosphotyrosine-containing peptides, while the SH3 domains can enhance this interaction and/or bind to other cellular components. Both the SH2 and SH3 domains of the human Crk protein are required for differentiation of PC12 cells. Thus, Crk has a role in an NGF-induced signaling pathway that involves activation of p21ras. Furthermore, three proteins of 118 kDa, 125 kDa, and 136 kDa which specifically bind to the Crk SH3 domain have been identified.Specifications
| Crk | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified |
| 22 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Bovine, Canine, Chicken, Frog, Human, Mouse, Rat | |
| Human Crk aa. 102 to 304 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title