Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CXCR4, Alexa Fluor 700, Clone: 44717, R&D Systems™

Supplier:  R&D Systems FAB173N025

 View more versions of this product

Catalog No. FAB173N025

This item is not returnable. View return policy

Description

Description

Host Species: Mouse
Clone: 44717
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: Mouse 3T3 cells transfected with human CXCR4.

Applications:

Flow Cytometry

Specifications

Specifications

CXCR4
Monoclonal
Alexa Fluor 700
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor
Mouse
Protein A or G purified from ascites
Primary
Reacts specifically with human and non-human cells expressing human CXCR4 (fusin) as detected by flow cytometry. It will also react with cells expressing feline CXCR4 but not rat CXCR4. This antibody does not cross-react with other chemokine receptors.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
IgG2b
Flow Cytometry
44717
Flow Cytometry 5 uL/10^6 cells
CXCR4
Mouse 3T3 cells transfected with human CXCR4
25 Tests
7852
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.