Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CXCR4, Alexa Fluor 700, Clone: 44717, R&D Systems™
Specifications
Antigen | CXCR4 |
---|---|
Clone | 44717 |
Dilution | Flow Cytometry 5 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB173N025
|
R&D Systems
FAB173N025 |
25 Tests | N/A | N/A | N/A | ||||
FAB173N100
|
R&D Systems
FAB173N100 |
100 Tests | N/A | N/A | N/A | ||||
Description
Host Species: Mouse
Clone: 44717
Isotype: IgG2B
Species Reactivity: Hu
Immunogen: Mouse 3T3 cells transfected with human CXCR4.
Applications:
Flow Cytometry
Specifications
CXCR4 | |
Flow Cytometry 5 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
7852 | |
Mouse 3T3 cells transfected with human CXCR4 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
44717 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201ESDF-1 receptor, FB22, Fusin, HM89NPYRL, HSY3RR, LAP3, LCR1, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3RWHIM, NPYR, NPYY3R, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor | |
CXCR4 | |
IgG2b | |
Protein A or G purified from ascites | |
Reacts specifically with human and non-human cells expressing human CXCR4 (fusin) as detected by flow cytometry. It will also react with cells expressing feline CXCR4 but not rat CXCR4. This antibody does not cross-react with other chemokine receptors. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title