Learn More
Cyclin B1 Mouse, Unlabeled, Clone: GNS-1, BD
Mouse Monoclonal Antibody
$342.00 - $406.00
Specifications
Antigen | Cyclin B1 |
---|---|
Clone | GNS-1 |
Concentration | 0.5mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Cyclins and cyclin-dependent kinases (cdks) are evolutionarily conserved proteins that are essential for cell-cycle control in eukaryotes. Cyclins (regulatory subunits) bind to cdks (catlytic subunits) to form complexes that regulate the progression of the cell cycle. The main cyclin-cdks complexes formed in vertebrate cells are cyclin D-cdk4 (G0/G1), cyclin E-cdk2 (G1/S), cyclin A-cdk2 (S) and cyclin B1-cdk1 (G2/M). These complexes are regulated by activating and inhibitory phosphorylation events, as well as by interactions with small regulatory proteins, such as p21 and p27 [Kip1]. Cyclin B1 is a mitotic cyclin, where expression is normally low in G0/G1, increases in S and is maximal during the G2/M phase. Cyclin B1 is rapidly degraded at the end of mitosis, and is required for cells to exit from mitosis. This antibody has been reported to react to hamster and mouse cyclin B1. In addition, the GNS-1 antibody has been reported to recognize an epitope between amino acids 1-21 of human cyclin B1.
Host Species: Mouse
Clone: GNS-1
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Cyclin B1 Recombinant Protein
Formula Weight [Chemical]: 62kDa
Bioimaging, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blotting
Specifications
Cyclin B1 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
GNS-1 | |
Western Blot | |
Unconjugated | |
RUO | |
Hamster, Human, Mouse | |
Human Cyclin B1 Recombinant Protein | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.