Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucovorin Calcium, USP, 95-105%, Spectrum™ Chemical
SDP

Catalog No. 18603794 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle

Catalog No. 18-603-794

Supplier: Spectrum Chemical Manufacturing Corporation LE117500GMBL

Only null left
Add to Cart
Add to Cart
Request a Quote

LE117, 1492-18-8, C20H21CaN7O7

Spectrum™ Chemical Leucovorin Calcium, USP is used to treat or prevent serious blood cell disorders caused by certain drugs that can reduce the effects of folic acid in the body. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1492-18-8
Molecular Formula C20H21CaN7O7
Molecular Weight (g/mol) 511.51
MDL Number MFCD00006704
InChI Key KVUAALJSMIVURS-AEQORCIRNA-L
IUPAC Name calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
SMILES [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 95 to 105%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.