Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DGKθ Mouse, Unlabeled, Clone: 24, BD
Mouse Monoclonal Antibody
Specifications
Antigen | DGKθ |
---|---|
Clone | 24 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB610931
|
BD Biosciences
610931 |
150 μg | N/A | N/A | N/A | ||||
Description
The protein kinase C pathway is a major signal transduction system that is activated upon stimulation of transmembrane receptors by hormones, neurotransmitters, and growth factors. Key mediators in this pathway are increased intracellular free Ca2+ levels and formation of diacylglycerol (DAG). DGKθ (diacylglycerol kinase θ) restricts PKC activation through the phosphorylation of DAG molecules that contain an unsaturated fatty acid at the sn-2 position to produce phosphatidic acid (PA). DGKθ contains several regions that are found in signaling molecules where they function in lipid-protein and protein-protein interactions. A C-terminal catalytic domain, three CRDs (cysteine rich domains), a PH domain, and an N-terminal proline/glycine rich domain are structural features of DGKθ. Six potential PKC phosphorylation sites lie between CRD3 and the PH domain. Cell-specific expression differentiate multiple isoforms of DGK. DGKθ is expressed primarily within the cerebellar cortex and hippocampus of the brain, but is also found in the small intestine and liver. The presence of the RA (Ras-associating) domain suggests that DGKθ may mediate activity of the Ras-like small GTP binding proteins.Specifications
DGKθ | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
24 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rabbit, Rat | |
Human DGKθ aa. 677-883 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title