Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Histone H3 (Lys4) Rabbit anti-Human, Mouse, Rat, Chicken, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$144.24 - $615.02

Specifications

Antigen dimethyl Histone H3 (Lys4)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Target Species Chicken, Human, Mouse, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABE250SMI
View Documents
MilliporeSigma
ABE250S
10 μL
Each for $144.24
Only null left
Add to Cart
 
ABE250MI
View Documents
MilliporeSigma
ABE250
100 μL
Each for $615.02
Only null left
Add to Cart
 
Description

Description

Anti-dimethyl Histone H3 (Lys4) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 dimethylated at lysine 4. This purified polyclonal antibody, also known as H3K4me2, has dot blot (DB) proven specificity and has been validated in western blot, immunocytochemistry, Chimmunoprecipitation.
Specifications

Specifications

dimethyl Histone H3 (Lys4)
Unconjugated
Chicken, Human, Mouse, Rat
NP_003484.1
KLH-conjugated linear peptide corresponding Histone H3 methylated at Lys4.
Affinity Purified
Polyclonal
Rabbit
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Primary
2-8°C for 12 months
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.