Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLP1 Mouse, Unlabeled, Clone: 8, BD

Catalog No. BDB611112 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611112 50 μg
BDB611113 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611112 Supplier BD Biosciences Supplier No. 611112
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dynamins, phosphoproteins with intrinsic GTPase activities, play an important role during early endocytosis. The GTPase activity is stimulated in vitro by microtubules, SH3 domain containing-proteins, and Protein Kinase (PKC)- mediated phosphorylation. Although highly related, dynamins are differentially expressed in an organism. For instance, dynamin I is found only in the nervous system, dynamin II is ubiquitously expressed, and dynamin III is located in lung, testis and brain. Due to four alternative splicing products for each of the dynamins, the dynamin family contains at least 12 different members. The Dynamin Like Protein-1 (DLP1) is a new member of the dynamin family of proteins. DLP1 (also known as DVLP) is widely expressed as two alternatively spliced products, each with different tissue expression patterns. The larger product is about 84kDa and reportedly found only in the brain, whereas the 79kDa product is ubiquitously expressed. Unlike dynamins I-III which co-localize with endocytotic vesicles, DLP1 is found aligned with microtubules and with the endoplasmic reticulum, suggesting a secretory function for this novel protein.

Host Species: Mouse
Clone: 8
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat DLP1 aa. 601-722
Formula Weight [Chemical]: 79-84kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen DLP1
Applications Western Blot
Classification Monoclonal
Clone 8
Concentration 250μg/mL
Conjugate Unconjugated
Description Dynamin Like Protein-1; DVLP
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat DLP1 aa. 601-722
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.