Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-DNA Polymerase iota, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB100175SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100175SS 0.025 mL
NB100175 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100175SS Supplier Novus Biologicals Supplier No. NB100175SS

Rabbit Polyclonal Antibody has been used in 4 publications

DNA Polymerase iota Polyclonal specifically detects DNA Polymerase iota in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen DNA Polymerase iota
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. Q9UNA4
Gene Alias DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB
Gene Symbols POLI
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human DNA Polymerase iota between amino acids 700-740, conjugated to KLH. [UniProt# Q9UNA4]
Molecular Weight of Antigen 80 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Polymerases, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 11201
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.