Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-DNA Polymerase iota, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
$220.00 - $342.00
Specifications
Antigen | DNA Polymerase iota |
---|---|
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100175SS
![]() |
Novus Biologicals
NB100175SS |
0.025 mL |
Each for $220.00
|
|
|||||
NB100175
![]() |
Novus Biologicals
NB100175 |
0.05 mL |
Each for $342.00
|
|
|||||
Description
DNA Polymerase iota Polyclonal specifically detects DNA Polymerase iota in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
DNA Polymerase iota | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Chromatin Research, DNA Polymerases, DNA Repair | |
Whole antisera with 0.02% Sodium Azide | |
DNA polymerase iota, EC 2.7.7.7, Eta2, polymerase (DNA directed) iota, RAD30 homolog B, RAD30Bpolymerase (DNA-directed), iota, RAD3OB | |
POLI | |
IgG | |
Unpurified | |
80 kDa |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse | |
Q9UNA4 | |
11201 | |
A synthetic peptide made to the C-terminal region of human DNA Polymerase iota between amino acids 700-740, conjugated to KLH. [UniProt# Q9UNA4] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title