Learn More
E-Cadherin Mouse, FITC, Clone: 36/E-CADHERIN, BD
Mouse Monoclonal Antibody
Specifications
Antigen | E-Cadherin |
---|---|
Clone | 36/E-CADHERIN |
Concentration | 250μg/mL |
Applications | Immunofluorescence |
Classification | Monoclonal |
Description
E-Cadherin is a 120kDa transmembrane glycoprotein that is localized in the adherens junctions of epithelial cells. There, it interacts with the cytoskeleton through the associated cytoplasmic catenin proteins. In addition to being a calcium-dependent adhesion molecule, E-Cadherin is also a critical regulator of epithelial junction formation. Its association with catenins is necessary for cell-cell adhesion. These E-cadherin/catenin complexes associate with corical actin bundles at both the zonula adherens and the lateral adhesion plaques. Tyrosine phosphorylation can disrupt these complexes, leading to changes in cell adhesion properties. E-Cadherin expression is often down-regulated in highly invasive, poorly differentiated carcinomas. Increased expression of E-Cadherin in these cells reduces invasiveness. Thus, loss of expression or function of E-Cadherin appears to be an important step in tumorigenic progression.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Specifications
E-Cadherin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
CD324; CDH1; CADH1; Cadherin-1; ECAD; CDHE; Arc-1; LCAM; UVO; Uvomorulin |
36/E-CADHERIN | |
Immunofluorescence | |
FITC | |
RUO | |
Canine, Human, Mouse, Rat | |
Human E-Cadherin aa. 735-883 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.