Learn More
eIF-4E Mouse, Unlabeled, Clone: 87, BD
Mouse Monoclonal Antibody
$287.50 - $574.50
Specifications
Antigen | eIF-4E |
---|---|
Clone | 87 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The eukaryotic translation initiation factor 4E (eIF-4E) is a 25kDa phosphoprotein that specifically binds to the 7-methylguanosine-containing cap of mRNA. eIF-4E is the rate-limiting component for the initiation of cap-dependent translation by the eIF-4E translation initiation complex. This complex promotes the unwinding of secondary structure at the 5′ untranslated region of mRNA, which is necessary to expose and locate the AUG-initiation codon. Phosphorylation of eIF-4E on Ser-209 occurs after serum treatment in CHO cells, and may regulate its function. Overexpression of eIF-4E can lead to increased cell proliferation, transformation, and tumorigenesis in nude mice. The overexpression of a Ala-53 variant of eIF-4E cannot evoke these changes, suggesting that Ser-53 on eIF-4E participates in the transfer of mRNA to the 48S initiation complexes. In cooperation with nuclear oncogenes such as c-myc or E1A, eIF-4E transforms primary cells. Other studies have demonstrated that overexpression of eIF-4E causes activation of Ras and leads to a transformed phenotype. Subsequent overexpression of GAP then causes reversion of this phenotype. The mechanism by which eIF-4E plays a role in transformation is not clear, but it is postulated that high levels of eIF-4E may lead to the translation of mRNAs that are normally translationally repressed.
Host Species: Mouse
Clone: 87
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Rabbit eIF-4E aa. 1-217
Immunofluorescence, Immunoprecipitation
Specifications
eIF-4E | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
87 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.