Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ERCC6L, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP213969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213969 Supplier Novus Biologicals Supplier No. NBP213969

Rabbit Polyclonal Antibody

ERCC6L Polyclonal specifically detects ERCC6L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ERCC6L
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias ATP-dependent helicase ERCC6-like, DNA excision repair protein ERCC-6-like, EC 3.6.1, EC 3.6.4.12, excision repair cross-complementing rodent repair deficiency, complementationgroup 6-like, excision repair protein ERCC6-like, FLJ20105, MGC131695, PICHexcision repair cross-complementing rodent repair deficiency complementationgroup 6 - like, PLK1-interacting checkpoint helicase, SNF2/RAD54 family protein, Tumor antigen BJ-HCC-15
Gene Symbols ERCC6L
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: IIWIRLVPLQEEIYRKFVSLDHIKELLMETRSPLAELGVLKKLCDHPRLLSARACCLLNLGTFSAQDGNEGEDSPDVDHIDQVTDDTLMEESGKMIFLMDLL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 54821
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.