Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ERK1, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB10056376SS

 View more versions of this product

Catalog No. NB10056376T


Description

Description

ERK1 Polyclonal specifically detects ERK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ERK1
Polyclonal
Unconjugated
PBS containing 0.2% gelatin with 0.05% Sodium Azide
EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24
Rabbit
Protein G purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
NP_001103361
MAPK3
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human ERK1 between amino acids 350-400.
0.025 mg
Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, Signal Transduction
5595
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only