Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ERK1, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$401.00

Specifications

Antigen ERK1
Concentration 0.5 mg/mL
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056376
SDP
View Documents
Novus Biologicals
NB10056376
0.1 mg
Each for $401.00
Only null left
Add to Cart
 
NB10056376T
SDP
View Documents
Novus Biologicals
NB10056376SS
0.025 mg N/A N/A N/A
Description

Description

ERK1 Polyclonal specifically detects ERK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ERK1
Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Polyclonal
Purified
RUO
Human
NP_001103361
5595
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human ERK1 between amino acids 350-400.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, Signal Transduction
PBS containing 0.2% gelatin with 0.05% Sodium Azide
EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24
MAPK3
IgG
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.