Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor), Mouse, Clone: 4F6-1D6, Abnova™

Catalog No. 89014107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-107 Supplier Abnova Corporation Supplier No. H00002170M01
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FABP3.

The intracellular fatty acid-binding proteins (FABPs) belongs to a multigene family. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Fatty acid-binding protein 3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is a candidate tumor suppressor gene for human breast cancer. [provided by RefSeq

Sequence: MVDAFLGTWKLVDSKNFDDYMKSLGVGFATRQVASMTKPTTIIEKNGDILTLRTHSTFKNTEISFKLGVEFDETTADDRKVKSIVTLDGGKLVHLQKWDGQETTLVRELIDGKLILTLTHGTAVCTRTYEKEA

Specifications

Antigen fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor)
Applications ELISA, KnockDown, Western Blot
Classification Monoclonal
Clone 4F6-1D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant FABP3.
Formulation PBS with no preservative; pH 7.4
Gene FABP3
Gene Accession No. BC007021
Gene Alias FABP11/H-FABP/MDGI/O-FABP
Gene Symbols FABP3
Host Species Mouse
Immunogen FABP3 (AAH07021, 1 a.a. ∼ 133 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 2170
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.