Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein (FITC) Mouse anti-All, Clone: 5D6.2, MilliporeSigma™

Catalog No. MAB045MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB045MI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB045MI Supplier MilliporeSigma Supplier No. MAB045
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Fluorescein (FITC) Antibody, clone 5D6.2 is validated for use in ELISA for the detection of Fluorescein (FITC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescein
Applications ELISA
Classification Monoclonal
Clone 5D6.2
Conjugate Unconjugated
Formulation Liquid in 0.02Mphosphate buffer, 0.25MNaCl, pH 7.6, 0.1% sodium azide.
Host Species Mouse
Immunogen Fluorescein-KLH
Quantity 1 mg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.