Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Fluorescein Isothiocyanate (FITC), Clone: F4/1, Epredia™ LabVision

Mouse Monoclonal Antibody

Specifications

Antigen Fluorescein Isothiocyanate (FITC) Ab-1
Classification Monoclonal
Conjugate Unconjugated
Form Purified
Host Species Mouse
View More Specs

Products 4
Catalog Number Mfr. No. Conjugate Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Conjugate Concentration Quantity Price Quantity & Availability  
MS207P0
View Documents
Epredia
MS207P0
Unconjugated 200μg/mL 100 μL N/A N/A N/A
MS207P1
View Documents
Epredia
MS207P1
Unconjugated 200μg/mL 500 μL N/A N/A N/A
MS207P
View Documents
Epredia
MS207P
Unconjugated 200μg/mL 1 mL N/A N/A N/A
MS207P1ABX
View Documents
Epredia
MS207P1ABX
Unconjugated 1mg/mL 100 μL N/A N/A N/A
Description

Description

Host Species: Mouse
Clone: F4/1
Isotype: IgG1/κ
Cross Reactivity: All species
Immunogen: Fluorescein isothiocyanate (FITC)

Ab-1 recognizes both the free and protein-conjugated (either soluble or cell bound) form of fluorescein or carboxyfluorescein. Sensitivity of the FITC-anti-FITC system is reportedly similar to that of the biotin-avidin system. In biotin or enzyme-labeled form, NeoMarkers' Ab-1 is also suitable for the conversion of fluorescein-labeled detection into immuno-enzymatic assays.

Flow Cytometry, Immunohistochemistry

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

Fluorescein Isothiocyanate (FITC) Ab-1
Unconjugated
Mouse
Fluorescein isothiocyanate (FITC)
Primary
Monoclonal
Purified
All species
IgG1 κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.