Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMC7 Mouse anti-Human, FITC, Clone: FMC7, BD

Catalog No. BDB340919 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340919 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340919 Supplier BD Biosciences Supplier No. 340919
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FMC7 recognizes a 105kDa membrane glycoprotein expressed on a subset of B lymphocytes.More than 50% of the peripheral B lymphocytes of normal adults carry the FMC7 antigen at variable density. FMC7-positive B cells are more mature, and they include the subpopulation that responds in vitro to mitogens or antigens. The FMC7 antigen is found on B-cell malignancies of most differentiated stages, such as mantle cell lymphoma, but not in most cases of chronic lymphocytic leukemia (CLL).

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen FMC7
Applications Flow Cytometry
Classification Monoclonal
Clone FMC7
Concentration 25μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human B-lymphoblastoid cell line HRIK
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.