Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMN2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP224615SS

 View more versions of this product

Catalog No. NBP224615SS


Only null left
Add to Cart

Description

Description

FMN2 Polyclonal specifically detects FMN2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FMN2
Polyclonal
Unconjugated
0.1 ml PBS with 0.05% Sodium Azide
FMN2
A synthetic peptide corresponding to amino acids 657-688 of human FMN2 was used as immunogen, Gene Bank no NP_064450.3.
0.025 mg
Signal Transduction
56776
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:100-1:4000, Immunohistochemistry-Paraffin 1:300-1:500
formin 2
Rabbit
Protein A purified
RUO
Primary
The amino acid sequence used as immunogen is 100% homologous in human and 77% homologous in mouse and rat.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only