Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FoxH1, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP145574 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145574 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145574 Supplier Novus Biologicals Supplier No. NBP145574

Specifications

Antigen FoxH1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Gene Accession No. O88621
Gene Alias Fast-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer
Gene Symbols FOXH1
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 55 of mouse FoxH1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8928
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.