Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G3BP Mouse anti-Human, Unlabeled, Clone: 23, BD

Catalog No. p-3970678 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611126 50 μg
BDB611127 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611126 Supplier BD Biosciences Supplier No. 611126
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Ras proteins are low molecular weight GTPases that play a critical role in the control of proliferation, differentiation, and cytoskeletal organization. RasGAP acts as a negative regulator of Ras, as well as a downstream target of Ras. It participates in the signal transduction cascade through interactions with other signaling proteins. Via its C-terminal domain, RasGAP interacts with Ras, but the N-terminus interacts with other signaling molecules such as p62 and p190. This function of RasGAP is mediated by the presence of SH2, SH3, and PH domains in the N-terminal region. However, some signaling proteins cannot bind to these domains. In an attempt to identify RasGAP-interacting molecules, a GAP SH3-Binding Protein (G3BP) was identified. This protein exhibits sequence homology to the hnRNP superfamily. It contains RNP1 and RNP2 motifs and is found primarily in the cytosol. G3BP interacts with GAP in proliferating cells and is dependent on the activation state of Ras. In addition, G3BP contains an intrinsic endonuclease activity that cleaves 3′-untranslated region (3′-UTR) mRNA. These data indicate that G3BP may link the Ras pathway to mRNA degradation.

Host Species: Mouse
Clone: 23
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human G3BP aa. 210-323
Formula Weight [Chemical]: 55-70kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen G3BP
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human G3BP aa. 210-323
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.