Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Galectin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa534913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534913 Supplier Invitrogen™ Supplier No. PA534913
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, LGALS4 transfected 293T lysate, Mouse colon, rat colon. Predicted reactivity: Dog (82%), Pig (84%), Rabbit (89%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The expression of this gene is restricted to small intestine, colon, and rectum, and it is underexpressed in colorectal cancer. Galectin 4 binds lactose and a related range of sugars. May be involved in the assembly of adherens junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Galectin 4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Lgals4
Gene Accession No. P38552, P56470, Q8K419
Gene Alias Antigen NY-CO-27; GAL4; gal-4; galectin 4; galectin-4; L-36; L-36 lactose-binding protein; L36LBI; L36LBP; lactose-binding lectin 4; Lectin galactose binding soluble 4 (Galectin-4); lectin, galactose binding, soluble 4; Lectin, galactose binding, soluble 4 (Galectin-4); lectin, galactoside binding soluble 4; lectin, galactoside-binding, soluble, 4; lectin, galactoside-binding, soluble, 4 (galectin 4); Lgals4
Gene Symbols Lgals4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Galectin 4. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16855, 25474, 3960
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.